Cresyl diphenyl phosphate (mixture of analogue) mixture of isomers ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Diphenyl p-tolyl phosphate, Diphenyl Cresyl Phosphate, Phosphoric Acid Cresyl Diphenyl Ester
CAS Number:26444-49-5
:
Formula:C₁₉H₁₇O₄P
MDL Number:MFCD00014920
Boiling Pt:390 °C
Melting Pt:–38 °C
Density:1.21
Molecular Weight:340.3097 g/mol
Flash Pt:232 °C (450 °F)
Storage Temperature:Ambient
EINECS:247-693-8
UN:3082
ADR:9 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together