Potassium-2-ethylhexanoate solution ≥75% (w/w)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Potassium 2-ethylhexanoate, 2-Ethylhexanoic Acid Potassium Salt
CAS Number:3164-85-0
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00045896
Molecular Weight:182.30 g/mol
Storage Temperature:Ambient
EINECS:221-625-7
UN:1993
ADR:3 III
:GHS02
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together