Anti-TOR3A Rabbit Polyclonal Antibody
Catalog # ABGEAP17612C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Torsin family 3, member A
- Antigen symbol:TOR3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Gene ID:64222
- Antigen synonyms:ADIR2|ADIR
- Amino acid number:187 to 213
- Storage buffer:pH 7,4
- Molecular weight:46.199 kDa
- Sequence:HFPHPKYVDLYKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.3
- Shipping temperature:4 °C
- Immunogen:This TOR3A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 187-213 amino acids from the Central region of human TOR3A.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TOR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human