Safety boots, lace-up, Rock & Roll, Cougar UK
Supplier: U-Power
Certificates
About this item
100% metal-free safety shoes made from pull-up water repellent leather with PU scuff and a slip-resistant dual-density PU sole.
- Comfortable, anatomically formed insole ergo dry features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
- Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
- Stress-out tongue system allows easy instep and provides added comfort when walking
- Natural fit width 11 for additional comfort
- Air toe protective aluminium toecap with breathable membrane (200 J)
- Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certifications: EN 20345:2011; S3, SRC
Frequently Bought Together