Laboratory air filtration system, HALO®
Supplier: ERLAB CAPTAIRE
New Product
Certificates
About this item
Ceiling mounted, laboratory grade filtration unit.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://ie.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=17633926
- Removes fugitive contaminants/odours in the laboratory
- 24/7 filtration
- Low cost - no duct work required
- Easy to install in the ceiling - no need to connect to the HVAC
- Easy to operate: The user can easily change the status, the setting, or check the routine maintenance via their smartphone
Halo continuously pulls dirty air in the laboratory through the filter, absorbing the molecules that cause noxious odours, and returning clean air back to the room through low disruptive side vents. The laboratory-grade molecular filtration system contains Neutrodine, a unique and powerful activated carbon mixture combined with a pre-filter that captures a large variety of airborne contaminants, which, left untreated, could represent a potential health hazard.
Caution: There is a very wide range of Erlab Fume Hoods and Enclosures. To ensure you receive the correct product for your specific application and setting please complete the contact form as per the url above. Your local Avantor sales representative will then send you an eValiQuest form to get details of your application and requirements. Once we have received this back from you, we will send you a quotation based on your precise requirements ensuring all handling and safety requirements are met.
Specifications
- Catalog No:
- ERLAAP026C09010031
- ERLAAP026C09020031
- Model:
- Halo 25 Smart white
- Halo 25 Smart black
- Lighting:
- LED
- LED
- Airflow:
- 110 m3/h (HEPA only 150 m3/h)
- 110 m3/h (HEPA only 150 m3/h)
- Energy consumption:
20 W (Bifiltration 35 W)
20 W (Bifiltration 35 W)
Specifications
Frequently Bought Together