Order Entry
Ireland
ContactUsLinkComponent
Anti-DERP6 Rabbit Polyclonal Antibody
Anti-DERP6 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9638R
Supplier:  Bioss
Anti-DERP6 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9638R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    DERP6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Antigen synonyms:
    Dermal papilla derived protein 6|HSPC002|DERP6|chromosome 17 open reading frame 81|MSTP071|MST071|ELP5_HUMAN.|S phase 2 protein|dermal papilla-derived protein 6
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

DERP6, is a 316 amino acid protein that localizes to the cytoplasm and exists as multiple alternatively spliced isoforms. Expressed ubiquitously with highest expression in liver, heart, testis, brain and skeletal muscle, DERP6 is thought to be involved in p53-mediated transcriptional regulation. The gene encoding DERP6 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DERP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat