Anti-ARHI Rabbit Polyclonal Antibody
Catalog # BOSSBS-2903R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ARHI
- Antigen symbol:ARHI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:9077
- Antigen synonyms:ARHI, NOEY2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:180-229/229
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
ARHI contains a highly conserved GTP-binding domain, a putative effector domain distinct from that of RAS and RAP proteins, and a C-terminal membrane localization motif. ARHI mRNA is detected in all normal breast and ovarian epithelial cell cultures tested, as well as in normal ovary, heart, liver, pancreas and brain however ARHI expression is down-regulated in breast and ovarian tumors. Reactivation of ARHI expression in breast cancer cells is associated with increased histone H3 acetylation and decreased lysine 9 methylation of histone H3.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ARHI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human