Anti-GPAT4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-15587R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:1 acyl sn glycerol 3 phosphate acyltransferase zeta
- Antigen symbol:GPAT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:LPAAT zeta|Glycerol-3-phosphate acyltransferase 4|GPAT4|Acyl-CoA:glycerol-3-phosphate acyltransferase 4|1-AGP acyltransferase 6|1-acylglycerol-3-phosphate O-acyltransferase 6|1-AGPAT 6|Lysophosphatidic acid acyltransferase zeta.|LPAAT-zeta|GPAT4_HUMAN|1 AGP acyltransferase 6|1 AGPAT 6|AGPAT6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.[supplied by OMIM, Apr 2004].
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: GPAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat