Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Nephrin Rabbit Polyclonal Antibody
Catalog # BOSSBS-10233R
Supplier:  Bioss
undefined
Anti-Nephrin Rabbit Polyclonal Antibody
Catalog # BOSSBS-10233R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nephrin
  • Antigen symbol:
    NPHS1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Gene ID:
    4868
  • Antigen synonyms:
    Renal glomerulus-specific cell adhesion receptor|NPHS1|NPHN|CNF|nephrin
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    480-530/1241
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Seems to play a role in the development or function of the kidney glomerular filtration barrier. Regulates glomerular vascular permeability. May anchor the podocyte slit diaphragm to the actin cytoskeleton. Plays a role in skeletal muscle formation through regulation of myoblast fusion (By similarity).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Nephrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat