Bottles, total organic carbon (TOC)
Supplier: EP SCIENTIFIC
Certificates
About this item
Bouston bottles are designed for the Total Organic Carbon (TOC) testing and sampling.
- USP method 643 testing
- Off-line and grab sampling of high purity water
- Low background is perfect for preparation and storage of standards
- Cleaning validation is simplified and costs are reduced by using the TOC method of testing
Specifications
- Catalog No:
- EPSC1000A/TOC
- EPSCS114-250C/TOC
- EPSCS114-250A/TOC
- EPSCS114-125A/TOC
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement