Anti-HEL308 Mouse Polyclonal Antibody
Catalog # ABNOH00113510-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:helicase, POLQ-like
- Antigen symbol:HELQ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Gene ID:113510
- Antigen synonyms:HEL308
- Amino acid number:1 to 272
- Sequence:MDECGSRIRRRVSLPKRNRPSLGCIFGAPTAAELEPGDEGKEEEEMVAENRRRKTAGVLPVEVQPLLLSDSPECLVLGGGDTNPDLLRHMPTDRGVGDQPNDSEVDMFGDYDSFTENSFIAQVDDLEQKYMQLPEHKKHATDFATENLCSESIKNKLSITTIGNLTELQTDKHTENQSGYEGVTIEPGADLLYDVPSSQAIYFENLQNSSNDLGDHSMKERDWKSSSHNTVNEELPHNCIEQPQQNDESSSKVRTSSDMNRRKSIKDHLKMP
- Immunogen:HEL308 (AAH11863.1, 1 a.a. ~ 272 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human HEL308 protein.
Type: Primary
Antigen: HEL308
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human