About this item
A variety of polyester and specialty materials for thermal transfer printing in wide format rolls designed for shape cutting multiple labels out of a single continuous roll.
B-423
This material is ideal for smooth-surface indoor labeling such as datacom equipment, patch panels, face plates, data ports and general industrial identification.
B-424
White matte paper labels with permanent rubber based adhesive for general purpose labelling.
B-428
Metallised polyester tape is ideal for indoor product labeling, asset tracking and general ID on smoother surfaces with a higher temperature limit.
B-432
Clear polyester tape is ideal for indoor labeling of smooth and slightly textured surfaces such as datacom equipment, faceplates, data ports, patch panels, and general industrial identification.
B-434 and B-486
Metallised polyester tape is ideal for indoor product labeling, asset tracking and general ID. B-434 is suitable for rough surfaces with a lower temperature limit. B-486 is suitable for very rough surfaces.
B-437
Topcoated polyvinylfluoride tape with a matte finish is designed for cable and wire bundle applications and label applications where self-extinguishing properties are required. Exhibits good solvent and heat resistance.
B-483
This material with a glossy finish is designed for high adhesion to textured metals and low surface energy plastics. For indoor use.
B-483/B-634
ToughStripe® floor marking features an ultra-aggressive adhesive which allows for printable messages to be placed on the floor. The floor marking roll is supplied with a separate roll of B-634 polyester overlaminate to protect your legend. Easy 3-step process: print label, position on floor, manually apply overlaminate. Withstands fork lift and pallet jack traffic without tearing or lifting.
B-484
Glossy white polyester tape with very aggressive adhesive ideal for angled and curved surface indoor labeling such as for panel labels and rating plates.