Order Entry
Ireland
ContactUsLinkComponent
 
Anti-mCherry Rabbit Polyclonal Antibody
  ROCK600-401-P16S
undefined
Anti-mCherry Rabbit Polyclonal Antibody
  ROCK600-401-P16S

 

  • Antigen symbol:
    DsRed
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Concentration:
    1,1 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    The immunogen is a mCherry mutant varient fusion protein of RFP corresponding to the full length amino acid sequence (234aa) derived from the mushroom polyp coral Discosoma.
  • Purification:
    mCherry was prepared from monospecific antiserum by imunoaffinity chromatography using Red Fluorescent Protein (Discosoma) coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities.   Expect reactivity against mCherry, RFP and its variants: tdTomato, mBanana, mOrange, mPlum, mOrange and mStrawberry. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum and purified and partially purified mCherry.  No reaction was observed against Human, Mouse or Rat serum proteins.  ELISA was used to confirm specificity at less than 0.1% of target signal.
  • Size:
    25 µl
  • Pk:
    25 µl

 

 

mCherry antibody is ideal for western blotting. Fluorescent proteins such as Discosoma Red Fluorescent Protein (and its varients), and GFP are widely used in research practice. Both commonly serve as a markers for gene expression and protein localization. DsRed was isolated from sea anemone Discosoma sp. mushroom and GFP is originated from Aequorea victoria jellyfish. As DsRed and GFP share only 19% identity, therefore, in general, anti-GFP antibodies do not recognize DsRed protein and vice versa. Structurally, Discosoma red fluorescent protein is similar to Aequorea green fluorescent protein in terms of its overall fold (a β-can) and chromophore-formation chemistry. However, Discosoma red fluorescent protein undergoes an additional steps in the chromophore maturation and obligates tetrameric structure. All mCherry antibodies have been pre-absorbed to eliminate any potential cross-reactivity to human, mouse and rat serum proteins. The antibodies are also confirmed for non-reactivity to GFP protein.

contains 0.01% (w/v) Sodium Azide

Type: Tag
Antigen: mCherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: