Anti-PMVK Antibody Pair
Catalog # ABNOH00010654-PW2
Supplier: Abnova
Specifications
- Antigen name:Phosphomevalonate Kinase
- Antigen symbol:PMVK
- Host:Multiple
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:10654
- Antigen synonyms:PMKASE|hPMK|PMKA|HUMPMKI|PMKI|PMVK|Phosphomevalonate kinase|PMK
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-PMVK (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-PMVK (50 µl)
Type:
Antigen: PMVK
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human