Anti-MCC Antibody Pair
Catalog # ABNOH00004163-AP51
Supplier: Abnova
Specifications
- Antigen name:Mutated in colorectal cancers
- Antigen symbol:MCC
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:4163
- Antigen synonyms:MCC-1|Mutated in colorectal cancers|CRCM_HUMAN.|MCC1|Colorectal mutant cancer protein|MCC 1|Protein MCC
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human MCC.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-MCC (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-MCC (50 µg);
Type:
Antigen: MCC
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human