Anti-CRYGD Polyclonal Antibody Pair
Catalog # ABNOH00001421-AP21
Supplier: Abnova
Specifications
- Antigen name:Crystallin, gamma D
- Antigen symbol:CRYGD
- Clonality:Polyclonal
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:1421
- Antigen synonyms:cry-g-D|CCA3|CCP|CTRCT4|CRYG4|PCC|CACA
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human CRYGD.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CRYGD (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-CRYGD (20 µg);
Type:
Antigen: CRYGD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human