Anti-LRG1 Antibody Pair
ABNOH00116844-AP11
: Abnova
- Antigen name:Leucine-rich alpha-2-glycoprotein 1
- Antigen symbol:LRG1
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:116844
- Antigen synonyms:leucine rich alpha 2 glycoprotein|LRG 1|1300008B03Rik|HMFT1766|2310031E04Rik|leucine-rich alpha-2-glycoprotein|LRG-1|LRG
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Set comes with matched Antibody Pair to detect and quantify protein level of human LRG1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-LRG1 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-LRG1, IgG2b kappa (20 µg);
Type:
Antigen: LRG1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human