- Description:Clean-Trace™ ATP Positive Control 3 ng
- Seq:03
- Environmentally Preferable:
- Pk:10
The Clean-Trace™ luminometer is a simple, quick and reliable handheld diagnostic tool to verify cleaning effectiveness and capture, store and manage data for later analysis. This Hygiene Monitoring and Management System offers food safety professionals a complete system of products consisting of a luminometer handheld device, compatible tests and a powerful new software platform, to verify that surfaces have been effectively cleaned in seconds, for better decision making and audit preparedness.
- Improved precision and accuracy
- Robust for the manufacturing environment
- Ergonomic design for simple one-handed operation
- Wireless data transfer
- 23 languages for multilingual pre-production teams
- Robust to provide consistent results across temperatures
The Clean-Trace™ luminometer is supplied with Clean-Trace™ Hygiene Monitoring and Managment Software with a newly redesigned dashboard for a quick and easy way to create, store and edit reports. The software turns data into information, helping you identify root causes and take action quickly. Now even easier to use and more intuitive, with convenient wireless and networking capabilities.