Anti-MS4A6A Rabbit Polyclonal Antibody
Catalog # ABNOPAB25749
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Membrane-spanning 4-Domains, Subfamily A, Member 6A
- Antigen symbol:MS4A6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:64231
- Antigen synonyms:4SPAN3|Membrane-spanning 4-domains subfamily A member 6A|CDA01|CD20L3|MST090|MSTP090|MS4A6A|CD20 antigen-like 3|4SPAN3.2|MS4A6
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 19 amino acids at N-terminus of human MS4A6A.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MS4A6A.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 2.5 ?g/ml
Type: Primary
Antigen: MS4A6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat