Anti-CTCF Rat Monoclonal Antibody [clone: KT70]
Catalog # ABNOMAB10146
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CCCTC-binding Factor
- Antigen symbol:CTCF
- Clonality:Monoclonal
- Clone:KT70
- Conjugation:Unconjugated
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:13018
- Antigen synonyms:transcriptional repressor CTCF|CTCFL paralog|11-zinc finger protein|11 zinc finger transcriptional repressor
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C for one month. For long term storage store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to C-terminus of mouse Ctcf.
- Purification:Protein G purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rat monoclonal antibody raised against recombinant Ctcf.
Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml
Type: Primary
Antigen: CTCF
Clonality: Monoclonal
Clone: KT70
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: