Anti-SLC45A3 Rabbit Polyclonal Antibody
ABNOPAB21057
: Abnova
- Antibody type:Primary
- Antigen name:Solute Carrier Family 45 Member A3
- Antigen symbol:SLC45A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:85414
- Antigen synonyms:IPCA6|PCANAP8|IPCA-2|IPCA-6|PCANAP6|PCANAP2|PRST|IPCA-8
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:PKYRGDTGGASSEDSLMTSFLPGPKPGAPFPNGHVGAGGSGLLPPPPALCGASACDVSVRVVVGEP
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human SLC45A3.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant SLC45A3.
Recommended Dilutions: Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: SLC45A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human