Anti-KCNA6 Rabbit Polyclonal Antibody
ABNOPAB10104
: Abnova
- Antibody type:Primary
- Antigen name:Potassium channel, voltage gated shaker related subfamily A, member 6
- Antigen symbol:KCNA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:64358
- Antigen synonyms:KV1.6|PPP1R96|HBK2
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH ) corresponding to rat Kcna6.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of Kcna6.
Recommended Dilutions: Western Blot: 5 to 10 ?g/ml
Type: Primary
Antigen: KCNA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat