Anti-EPM2A Mouse Monoclonal Antibody [clone: k2A3]
Catalog # ABNOMAB2052
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Epilepsy progressive myoclonus type 2A Lafora disease (laforin)
- Antigen symbol:EPM2A
- Clonality:Monoclonal
- Clone:k2A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:7957
- Antigen synonyms:LAFPTPase|Laforin|Lafora PTPase|EPM2|EPM2A|MELF
- Amino acid number:243 to 331
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 243-331 of human EPM2A.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant EPM2A.
Type: Primary
Antigen: EPM2A
Clonality: Monoclonal
Clone: k2A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human