Anti-PLIN5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12159
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Perilipin 5
- Antigen symbol:PLIN5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:66968
- Antigen synonyms:MLDP|PAT-1|Lsdp5|2310076L09Rik|AW109675|AI415325
- Amino acid number:400 to 463
- Storage buffer:PBS (30% glycerol, 0,1% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 400-463 of mouse Plin5.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Plin5.
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml
Type: Primary
Antigen: PLIN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse