Anti-FN1 Mouse Monoclonal Antibody [clone: Fn-3]
Catalog # ABNOMAB7201
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fibronectin
- Antigen symbol:FN1
- Clonality:Monoclonal
- Clone:Fn-3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:2335
- Antigen synonyms:Cold insoluble globulin|ED-B|FNZ|MSF|CIG|GFND2|LETS|fibronectin 1|GFND|FN|FINC
- Storage buffer:PBS
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human FN1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant FN1.
Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: Fn-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human