Order Entry
Ireland
ContactUsLinkComponent
Anti-PARP4 Monoclonal Antibody [clone: P193-10]
Anti-PARP4 Monoclonal Antibody [clone: P193-10]
Catalog # ABNOMAB5188
Supplier:  Abnova
Anti-PARP4 Monoclonal Antibody [clone: P193-10]
Catalog # ABNOMAB5188
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Poly (ADP-ribose) polymerase 4
  • Antigen symbol:
    PARP4
  • Clonality:
    Monoclonal
  • Clone:
    P193-10
  • Conjugation:
    Unconjugated
  • Host:
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Amino acid number:
    408 to 611
  • Storage buffer:
    buffer containing 1% BSA, 0,1% sodium azide
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 408-611 of PARP4.
  • Size:
    1 mL
  • Pk:
    1 ml

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant PARP4.

Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:10-1:20

Type: Primary
Antigen: PARP4
Clonality: Monoclonal
Clone: P193-10
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b kappa
Reactivity: Human