Anti-TFB1M Goat Polyclonal Antibody
Catalog # ABNOPAB16639
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor B1, mitochondrial
- Antigen symbol:TFB1M
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:51106
- Antigen synonyms:homolog of yeast mitochondrial transcription factor B|N'-adenosyl(rRNA) dimethyltransferase 1|mitochondrial 12S rRNA dimethylase 1|S-adenosylmethionine-6-N'|mtTFB|CGI-75|mitochondrial|CGI75|transcription factor B1|OTTHUMP00000017473|mtTFB1|TFB1M
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-ELKRRKSKNEEKE
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human TFB1M.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of TFB1M.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: TFB1M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human