Order Entry
Ireland
ContactUsLinkComponent
Anti-CML Monoclonal Antibody (Peroxidase) [clone: CMS-10]
Anti-CML Monoclonal Antibody (Peroxidase) [clone: CMS-10]
Catalog # ABNOMAB6591
Supplier:  Abnova
Anti-CML Monoclonal Antibody (Peroxidase) [clone: CMS-10]
Catalog # ABNOMAB6591
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CML
  • Clonality:
    Monoclonal
  • Clone:
    CMS-10
  • Conjugation:
    Peroxidase
  • ELISA:
    Yes
  • Host:
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    2% Block Ace, 0,1% proclin
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein (conjugated with KLH) corresponding to CML.
  • Size:
    50 μg
  • Pk:
    50 µg

Specifications

About this item

Mouse monoclonal antibody raised against recombinant CML.

Recommended Dilutions: ELISA: 0.1-1 ?g/ml; Immunohistochemistry: 5-10 ?g/ml

Type: Primary
Antigen: CML
Clonality: Monoclonal
Clone: CMS-10
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse, Rat