Anti-EB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ABNOPAB14213
Supplier: Abnova
Specifications
- Antigen name:Elementary Bodies
- Antigen symbol:EB
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:C. trachomatis
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (10 mg/mL BSA, 0,002% thimerosal)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:L2 and other serovar groups.
- Pk:1 mL
Specifications
About this item
Rabbit polyclonal antibody raised against Chlamydia trachomatis, Elementary Bodies.
Type:
Antigen: EB
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: C. trachomatis