Anti-DNAJB11 Rabbit Polyclonal Antibody
Catalog # ABNOPAB17700
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DnaJ (Hsp40) homolog, subfamily B, member 11
- Antigen symbol:DNAJB11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:51726
- Antigen synonyms:DJ9|EDJ|ERj3|ABBP-2|ERdj3|PRO1080|Dj-9|ERj3p|ABBP2|UNQ537
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human DNAJB11.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of DNAJB11.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; ELISA: 1:20000
Type: Primary
Antigen: DNAJB11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat