Anti-TH1L Goat Polyclonal Antibody
Catalog # ABNOPAB18951
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Negative elongation factor complex member C/D
- Antigen symbol:NELFCD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center
- Cross adsorption:No
- Format:Liquid
- Gene ID:51497
- Antigen synonyms:NELF-D|TH1|NELF-C|HSPC130|TH1L
- Storage buffer:0,5 mg/mL Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-NKRVSINKDE
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at internal region of human TH1L.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of TH1L.
Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.5-2 ?g/ml
Type: Primary
Antigen: TH1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human