Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Listeria monocytogenes ActA Mouse Monoclonal Antibody [clone: 3a15]
  ABNOMAB8953
 :  Abnova
Anti-Listeria monocytogenes ActA Mouse Monoclonal Antibody [clone: 3a15]
  ABNOMAB8953
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Listeria monocytogenes ActA
  • Clonality:
    Monoclonal
  • Clone:
    3a15
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Amino acid number:
    117 to 278
  • Storage buffer:
    citrate-Tris-HCl, pH7.0 (0,02% Procl300)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 117-278 of Listeria monocytogenes ActA.
  • Purification:
    Affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Mouse monoclonal antibody raised against partial recombinant Listeria monocytogenes ActA.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Listeria monocytogenes ActA
Clonality: Monoclonal
Clone: 3a15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: