Anti-AHI1 Mouse Monoclonal Antibody [clone: 645s3]
Catalog # ABNOMAB8962
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Abelson helper integration site 1
- Antigen symbol:AHI1
- Clonality:Monoclonal
- Clone:645s3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:52906
- Antigen synonyms:ORF1|AHI-1|dJ71N10.1|JBTS3
- Storage buffer:PBS, pH 7,4 (0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to C-terminus of mouse Ahi-1.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant Ahi1.
Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunoprecipitation: 1-2 ?g/200-800 ?g protein; Immunohistochemistry: 1:200-1:1000 The optimal working dilution sho?ld be determined.
Type: Primary
Antigen: AHI1
Clonality: Monoclonal
Clone: 645s3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse