Order Entry
Ireland
ContactUsLinkComponent
Anti-LMTK2 Goat Polyclonal Antibody
Anti-LMTK2 Goat Polyclonal Antibody
  ABNOPAB19059
 :  Abnova
Anti-LMTK2 Goat Polyclonal Antibody
  ABNOPAB19059
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    lemur tyrosine kinase 2
  • Antigen symbol:
    LMTK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    internal
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    22853
  • Antigen synonyms:
    LMR2|KPI-2|BREK|AATYK2|cprk|PPP1R100|KPI2
  • Storage buffer:
    Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
  • Sequence:
    RCINSESSTD
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids at internal region of human LMTK2.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Goat polyclonal antibody raised against synthetic peptide of LMTK2.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: LMTK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human