Anti-Yc2 Goat Polyclonal Antibody
Catalog # ABNOPAB19635
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glutathione S-transferase alpha 3
- Antigen symbol:GSTA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:494500
- Antigen synonyms:Yc2|Gsta5
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-EMDPGIVDNFPLLK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region of rat Yc2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of Yc2.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: Yc2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat