Anti-Vtg Mouse Monoclonal Antibody [clone: JE-8D6]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: JE-8D6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Carp, fathead Minnow, Roach, Zebrafish
Specifications
- Catalog No:
- CAYM170166-100
- CAYM170166-500
- Antigen symbol:
- VTG
- VTG
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- JE-8D6
- JE-8D6
- Reactivity:
- Zebrafish, Carp, Fathead minnow, Roach
- Zebrafish, Carp, Fathead minnow, Roach
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Vtg from whole body homogenate of 17-beta-estradiol treated zebrafish (D. rerio)
- Vtg from whole body homogenate of 17-beta-estradiol treated zebrafish (D. rerio)
- Purification:
- purified
- purified
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together