Anti-TPH Sheep Polyclonal Antibody
Catalog # CAYM10009396-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:TPH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:recombinant rabbit TPH
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: TPH
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Rat