Anti-COUP-TF1 Rabbit Polyclonal Antibody
PIEAPA521611
- Antibody type:Primary
- Antigen name:Nuclear Receptor Subfamily 2, Group F, Member 1
- Antigen symbol:NR2F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:7025
- Antigen synonyms:EAR-3|v-erb-a homolog-like 3)|V-erbA-related protein 3|COUP transcription factor 1|EAR3|transcription factor COUP 1 (chicken ovalbumin upstream promoter 1|ERBAL3|TCFCOUP1|SVP44|COUP-TF I|TFCOUP1|COUP transcription factor I|COUP-TF1|chicken ovalbumin upstream promoter-transcription factor I|NR2F2|COUP-TFI
- Storage buffer:PBS, 10% glycerol, 0.01% thimerosal, pH 7
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide corresponding to a region within amino acids 1 and 44 of COUP TF1 (Uniprot ID#P10589)
- Purification:Antigen affinity chromatography
- Size:100 µL
- Pk:100 µG
Type: Primary
Antigen: COUP-TF1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human