Anti-RBL2 Rabbit Polyclonal Antibody
ROCK600-401-B11S
- Antibody type:Primary
- Antigen name:Retinoblastoma-like 2
- Antigen symbol:RBL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Chimpanzee,Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:5934
- Antigen synonyms:Rb2|P130
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,1% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared by repeated immunizations with a synthetic peptide corresponding to the Spa310 sequence of pRb2/p130 Protein. A residue of cysteine was added to facilitate coupling.
- Size:25 µl
- Pk:25 µl
This affinity purified antibody is suitable for ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 130 kDa in size corresponding to pRb2/p130, and a band approximately 4.2kDa.
Recommended Dilutions: ELISA: 1:75000 - 1:100000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: RBL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee