Anti-POGZ Rabbit Polyclonal Antibody
ROCK600-401-A89S
- Antibody type:Primary
- Antigen name:Pogo Transposable Element With ZNF Domain
- Antigen symbol:POGZ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Chicken,Macaque,Chimpanzee,Human,Bovine,Dog,Olive baboon,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:near the N-terminus
- Cross adsorption:No
- Gene ID:229584
- Antigen synonyms:9530006B08Rik
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the N-terminus of mouse Pogz protein.
- Purification:purified by immunoaffinity chromatography
- Size:25 µl
- Pk:25 µl
Recommended Dilutions: ELISA: 1:100000 to 1:400000; Western Blot: 1:2000 to 1:20000; Other Dilution: User Optimized
Type: Primary
Antigen: POGZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Short-Tailed Opossum, Bovine, Rat, Chimpanzee, Macaque, Olive Baboon, Chicken, Mouse