Anti-BIRA Rabbit Polyclonal Antibody
Catalog # ROCK100-401-B21
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen symbol:BIRA
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Virus
- Western blot:Yes
- Environmentally Preferable:
- ChIP:Yes
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: BIRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: