Anti-Lysosome Associated Membrane Protein 1 Mouse Monoclonal Antibody [clone: 4E151]
USBIL9190-10-200
- Antibody type:Primary
- Antigen name:Lysosome Associated Membrane Protein 1
- Antigen symbol:LAMP1
- Clonality:Monoclonal
- Clone:4E151
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Hamster,Human,Rat
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide, 50% glycerol.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Rat liver lysosomal membrane preparations.
- Purification:purified by Protein G chromatography
- Size:200 μg
- Pk:200 µG
Recommended dilution: Immunochemistry: 1:100
Type: Primary
Antigen: Lysosome Associated Membrane Protein 1
Clonality: Monoclonal
Clone: 4E151
Conjugation: unconjugated
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Hamster, Human, Rat,