Anti-Urokinase Plasminogen Activator Receptor Mouse Monoclonal Antibody [clone: VIM5]
Catalog # USBIU2610-03F-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Urokinase Plasminogen Activator Receptor
- Antigen symbol:uPA
- Clonality:Monoclonal
- Clone:VIM5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:Human Urokinase Plasminogen Activator Receptor
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Urokinase Plasminogen Activator Receptor
Clonality: Monoclonal
Clone: VIM5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,