Order Entry
Ireland
ContactUsLinkComponent
Anti-pro-Guanylin Mouse Monoclonal Antibody [clone: MAK-L-G-11]
Anti-pro-Guanylin Mouse Monoclonal Antibody [clone: MAK-L-G-11]
Catalog # USBIG9805-04E-100
Supplier:  US Biological
Anti-pro-Guanylin Mouse Monoclonal Antibody [clone: MAK-L-G-11]
Catalog # USBIG9805-04E-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    pro-Guanylin
  • Clonality:
    Monoclonal
  • Clone:
    MAK-L-G-11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    lyophilized powder
  • Amino acid number:
    78 to 94
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Synthetic human pro-Guanylin (aa 78-94)
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: pro-Guanylin
Clonality: Monoclonal
Clone: MAK-L-G-11

Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: