Order Entry
Ireland
ContactUsLinkComponent
Anti-Heat Shock Protein 90, beta Mouse Monoclonal Antibody [clone: 7L19]
Anti-Heat Shock Protein 90, beta Mouse Monoclonal Antibody [clone: 7L19]
Supplier:  US Biological
Certificates
Anti-Heat Shock Protein 90, beta Mouse Monoclonal Antibody [clone: 7L19]
Supplier:  US Biological

Specifications

  • Catalog No:
  • USBIH1834-55M-25
  • USBIH1834-55M-100
  • Antigen symbol:
  • HSP90B
  • HSP90B
  • Antigen name:
  • Heat Shock Protein 90 beta
  • Heat Shock Protein 90 beta
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 7L19
  • 7L19
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgM
  • IgM
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant corresponding to human Heat Shock Protein 90, beta
  • Recombinant corresponding to human Heat Shock Protein 90, beta
  • Form:
  • liquid
  • liquid
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Concentration:
  • ~1 mg/ml
  • ~1 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,2, 0,09% sodium azide, 50% glycerol.
  • PBS, pH 7,2, 0,09% sodium azide, 50% glycerol.
  • Storage temperature:
  • –20 °C
  • –20 °C

Specifications

Related Information

Frequently Bought Together