Anti-MAFB Mouse Monoclonal Antibody [clone: 2A6]
Catalog # ORIGCF800291
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:MAFB
- Clonality:Monoclonal
- Clone:2A6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:32 to 320
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35.6 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 32-320 of human MAFB (NP_005452) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: MAFB
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human