Order Entry
Ireland
ContactUsLinkComponent
 
DL-α-Tocopherol (Vitamin E), EMPROVE® ESSENTIAL USP for oral use, SAFC®
 
DL-α-Tocopherol (Vitamin E), EMPROVE® ESSENTIAL USP for oral use, SAFC®
 

 

  • Environmentally Preferable:

 

 

  • Assay of DL-α-tocopherol acetate:
    475 - 525 mg/ml
  • Identity:
    passes test
  • Appearance:
    clear, slightly yellow, viscous liquid
  • Acidity:
    passes test
  • Miscibility with water:
    passes test
  • pH-value (10 %; water):
    6.5 - 7.5
  • Density (d 20 °C/ 4 °C):
    about 1.01
  • Residual solvents (Ph. Eur./ICH):
    excluded by production process
  • Microbiological test:
    passes test

 

 

Frequently Bought Together