Order Entry
Ireland
ContactUsLinkComponent
Anti-1,25-dihydroxy-Vitamin D3 Mouse Monoclonal Antibody [clone: ID1,25A]
Anti-1,25-dihydroxy-Vitamin D3 Mouse Monoclonal Antibody [clone: ID1,25A]
Catalog # USBIV2129
Supplier:  US Biological
Anti-1,25-dihydroxy-Vitamin D3 Mouse Monoclonal Antibody [clone: ID1,25A]
Catalog # USBIV2129
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    1,25 Dihydroxy Vitamin D3
  • Clonality:
    Monoclonal
  • Clone:
    ID1,25A
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
  • Environmentally Preferable:
  • Form:
    lyophilized powder
  • Storage buffer:
    50mM Tris, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml (after reconstutution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    10 µg
  • Pk:
    10 µG

Specifications

About this item

Recommended dilution: ELISA: 1:25000

Type: Primary
Antigen: 1,25 Dihydroxy Vitamin D3
Clonality: monoclonal
Clone: ID1,25A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: