Anti-Synuclein, alpha Sheep Polyclonal Antibody
Catalog # USBIS9500-03L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Synuclein alpha
- Antigen symbol:SNCA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:
- Reactivity:Rodent,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:sterile water.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: Synuclein, alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Mouse, Rat, Rodent