Order Entry
Ireland
ContactUsLinkComponent
Anti-SMARCA4 Mouse Monoclonal Antibody [clone: BRG-01]
Anti-SMARCA4 Mouse Monoclonal Antibody [clone: BRG-01]
Catalog # USBIS1014-58Y7
Supplier:  US Biological
undefined
Anti-SMARCA4 Mouse Monoclonal Antibody [clone: BRG-01]
Catalog # USBIS1014-58Y7
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
  • Antigen symbol:
    Transcription activator BRG1
  • Clonality:
    Monoclonal
  • Clone:
    BRG-01
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 10 ?g/ml; Western Blot: 10 ?g/ml

Type: Primary
Antigen: SMARCA4 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4)
Clonality: Monoclonal
Clone: BRG-01
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human